Assay Details
Gene Symbol: GABRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000809.3 | NP_000800.2 | 9 | 9004 | 88 | |
NM_001204266.1 | NP_001191195.1 | 9 | 8034 | 88 | ||
NM_001204267.1 | NP_001191196.1 | 8 | 7881 | 88 | ||
GenBank mRNA | AF238869.1 | - | 1 | 4898 | 88 | |
BC036474.1 | - | 1 | 530 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
