Assay Details
Gene Symbol: LINC01021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CQ819099.1 | - | 3 - 4 | 472 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the CQ819099.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000503107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000505775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000510165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000512067.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | ENST00000514844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007000.hg.1 | NR_038848.1 | Refseq |
