Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000322611.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
ENST00000616743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_001271606.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013155.hg.1 |
NM_006317.4 |
Refseq |