Assay Details
Gene Symbol: LOC101927488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AW665322.1 | - | 2 - 3 | 479 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
