Assay Details
Gene Symbol: RIOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159749.1 | NP_001153221.1 | 1 - 2 | 179 | 157 | |
NM_018343.2 | NP_060813.2 | 1 - 2 | 179 | 157 | ||
GenBank mRNA | AK002021.1 | - | 1 - 2 | 118 | 157 | |
AK225348.1 | - | 1 - 2 | 100 | 157 | ||
BC000953.2 | - | 1 - 2 | 107 | 3449997 | 157 | |
DB240299.1 | - | 1 - 2 | 179 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
