Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122853.2 | NP_001116325.1 | 5 - 6 | 1011 | 81 | |
NM_133371.4 | NP_588612.2 | 5 - 6 | 663 | 81 | ||
GenBank mRNA | AF480443.1 | - | 5 - 6 | 515 | 81 | |
AJ300586.1 | - | 5 - 6 | 626 | 81 | ||
AJ300587.1 | - | 5 - 6 | 506 | 81 | ||
AK313873.1 | - | 5 - 6 | 667 | 81 | ||
BC069380.1 | - | 5 - 6 | 493 | 7262195 | 81 | |
BC069553.1 | - | 5 - 6 | 493 | 7262207 | 81 | |
BC074870.2 | - | 5 - 6 | 449 | 30915232 | 81 | |
BC074871.2 | - | 5 - 6 | 449 | 30915469 | 81 | |
BP360917.1 | - | 1 - 2 | 147 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000297130.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000456566.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517768.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000517861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000520112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000521300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000523553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000524256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | ENST00000615557.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_001122853.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009098.hg.1 | NM_133371.4 | Refseq |
