Assay Details
Gene Symbol: TNFAIP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286815.1 | NP_001273744.1 | 2 - 3 | 328 | 76 | |
NM_001286817.1 | NP_001273746.1 | 2 - 3 | 332 | 76 | ||
GenBank mRNA | DB114867.1 | - | 2 - 3 | 266 | 76 | |
DB124282.1 | - | 2 - 3 | 169 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286815.1, NM_001286817.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
