Assay Details
Gene Symbol: LOC643201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036494.1 | 8 - 9 | 964 | 75 | ||
GenBank mRNA | BC034407.1 | - | 8 - 9 | 1029 | 75 | |
BC052945.1 | - | 5 - 6 | 308 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
