Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000499625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000525963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007073.hg.1 |
ENST00000528701.1 |
Ensembl |