Assay Details
Gene Symbol: LOC101928540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125859.1 | 7 - 8 | 583 | 72 | ||
GenBank mRNA | AI809145.1 | - | 2 - 3 | 72 | ||
BX110992.1 | - | 2 - 3 | 230 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
