Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
ENST00000370693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008350.hg.1 |
NM_004282.3 |
Refseq |