Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000311378.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
ENST00000426127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008271.hg.1 |
NM_175575.5 |
Refseq |