Assay Details
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271844.1 | NP_001258773.1 | 8 | 6979 | 73 | |
NM_001271845.1 | NP_001258774.1 | 8 | 7018 | 73 | ||
NM_019020.3 | NP_061893.2 | 12 | 8134 | 73 | ||
XM_005257049.2 | XP_005257106.1 | 12 | 8145 | 73 | ||
XM_005257050.3 | XP_005257107.1 | 12 | 8148 | 73 | ||
XM_006721694.3 | XP_006721757.1 | 13 | 7987 | 73 | ||
XM_011524322.2 | XP_011522624.1 | 12 | 8109 | 73 | ||
XM_011524323.2 | XP_011522625.2 | 10 | 8059 | 73 | ||
XM_011524324.2 | XP_011522626.1 | 8 | 7603 | 73 | ||
XM_017024186.1 | XP_016879675.1 | 10 | 7886 | 73 | ||
XM_017024187.1 | XP_016879676.1 | 11 | 8126 | 73 | ||
XM_017024188.1 | XP_016879677.1 | 10 | 7266 | 73 | ||
GenBank mRNA | AL512709.1 | - | 1 | 3374 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
