Assay Details
Gene Symbol: CYP39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017010921.1 | XP_016866410.1 | 10 - 11 | 1473 | 76 | |
XM_017010922.1 | XP_016866411.1 | 10 - 11 | 1457 | 76 | ||
XM_017010923.1 | XP_016866412.1 | 9 - 10 | 1365 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
