Assay Details
Gene Symbol: LOC101927668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CK904648.1 | - | 5 - 6 | 523 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000415499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000418442.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000424516.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000435968.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000437191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000449573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | ENST00000457921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013523.hg.1 | NR_110114.1 | Refseq |
