Assay Details
Gene Symbol: JHDM1D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024451.1 | 1 | 1916 | 88 | ||
GenBank mRNA | AK026372.1 | - | 1 | 1822 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
