Assay Details
Gene Symbol: PKIA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BU161278.1 | - | 4 - 5 | 352 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000519242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522302.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | ENST00000522807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | NR_125389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010878.hg.1 | NR_125389.1 | Refseq |
