Assay Details
Gene Symbol: LOC101929563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AW900513.1 | - | 1 - 2 | 139 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | ENST00000440888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009739.hg.1 | NR_121602.1 | Refseq |
