Assay Details
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017019178.1 | XP_016874667.1 | 1 - 2 | 1668 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019178.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
