Assay Details
Gene Symbol: TTC41P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC033128.1 | - | 1 - 2 | 331 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000388789.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548520.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548527.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000548897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551270.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000551885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | ENST00000552065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011749.hg.1 | NR_027249.1 | Refseq |
