Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000312143.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000412676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000549466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000550573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
ENST00000552995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142343.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142344.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_001142345.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011838.hg.1 |
NM_004072.2 |
Refseq |