Assay Details
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003806.2 | NP_003797.1 | 1 | 243 | 58 | |
NR_073189.1 | 1 | 243 | 58 | |||
XM_017020146.1 | XP_016875635.1 | 1 | 239 | 58 | ||
GenBank mRNA | U76376.1 | - | 1 | 243 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
