Assay Details
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018638.4 | NP_061108.2 | 8 | 2727 | 84 | |
GenBank mRNA | CR749601.1 | - | 1 | 314 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
