Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004943.1 | NP_004934.1 | 3 - 4 | 1949 | 69 | |
GenBank mRNA | AK309497.1 | - | 1 - 2 | 67 | 69 | |
CD106470.1 | - | 1 - 2 | 602 | 69 | ||
L19267.1 | - | 3 - 4 | 1540 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
