Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |