Assay Details
Gene Symbol: C1QTNF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC144450.1 | - | 1 - 2 | 236 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382140.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000382145.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | ENST00000556521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NM_001007537.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008342.hg.1 | NR_104426.1 | Refseq |
