Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005267919.3 | XP_005267976.1 | 1 - 2 | 78 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267919.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
