Assay Details
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045529.3 | 3 - 4 | 575 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000613213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000614978.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008088.hg.1 | ENST00000620003.1 | Ensembl |
