Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166340.1 | NP_001159812.1 | 2 | 860 | 88 | |
NM_001172109.1 | NP_001165580.1 | 3 | 1014 | 88 | ||
NM_001172110.1 | NP_001165581.1 | 3 | 856 | 88 | ||
NM_001172111.1 | NP_001165582.1 | 3 | 873 | 88 | ||
NM_145204.3 | NP_660205.3 | 2 | 749 | 88 | ||
XM_005254157.3 | XP_005254214.1 | 2 | 629 | 88 | ||
XM_011521215.2 | XP_011519517.1 | 3 | 1147 | 88 | ||
XM_011521217.2 | XP_011519519.1 | 2 | 737 | 88 | ||
GenBank mRNA | AF308450.1 | - | 2 | 653 | 88 | |
AK097557.1 | - | 2 | 860 | 88 | ||
AK292298.1 | - | 2 | 751 | 88 | ||
AY008293.1 | - | 2 | 713 | 88 | ||
BC031411.1 | - | 2 | 626 | 5195120 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500009950.hg.1 | ENST00000568984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009950.hg.1 | ENST00000568984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009950.hg.1 | ENST00000568984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009950.hg.1 | ENST00000568984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
