Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000371244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
ENST00000610361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008450.hg.1 |
NM_006252.3 |
Refseq |