Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000499560.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011652.hg.1 |
ENST00000526611.1 |
Ensembl |