Assay Details
Gene Symbol: AMZ2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026903.1 | 1 - 2 | 1031 | 104 | ||
GenBank mRNA | AK056627.1 | - | 1 - 2 | 1014 | 104 | |
BX537538.1 | - | 1 - 2 | 905 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK056627.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
