Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000630622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
ENST00000630622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000335793.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000439510.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000578516.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000579611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000580315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000581045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000582303.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000630622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
ENST00000630622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011657.hg.1 |
NM_012121.4 |
Refseq |