Assay Details
Gene Symbol: FAM71E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145402.1 | NP_001138874.1 | 6 - 7 | 949 | 94 | |
GenBank mRNA | AK127979.1 | - | 5 - 6 | 871 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
