Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
ENST00000369448.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009572.hg.1 |
NM_017709.3 |
Refseq |