Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |