Assay Details
Gene Symbol: PPM1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC064628.1 | - | 5 - 6 | 889 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000309276.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000464951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000471106.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000482367.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | ENST00000486709.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018464.hg.1 | NM_005167.5 | Refseq |
