Assay Details
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024575.4 | NP_078851.2 | 2 | 252 | 131 | |
GenBank mRNA | AF271774.1 | - | 2 | 233 | 131 | |
AF548003.1 | - | 1 | 126 | 131 | ||
AK027120.1 | - | 2 | 228 | 131 | ||
AK225472.1 | - | 2 | 227 | 131 | ||
AL833373.1 | - | 1 | 2174 | 131 | ||
BC063014.1 | - | 2 | 204 | 5224593 | 131 | |
BI833715.1 | - | 2 | 252 | 131 | ||
CR457375.1 | - | 1 | 126 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
