Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540275.2 | XP_011538577.1 | 1 - 2 | 347 | 75 | |
GenBank mRNA | CR749429.1 | - | 1 - 2 | 314 | 75 | |
DB455338.1 | - | 1 - 2 | 376 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540275.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373878.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000373880.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000472199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | ENST00000486074.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_001143774.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007714.hg.1 | NM_032439.3 | Refseq |
