Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008010.hg.1 |
NM_006779.3 |
Refseq |