Assay Details
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ471858.1 | - | 1 | 118 | 103 | |
DQ471859.1 | - | 1 | 83 | 103 | ||
DQ471860.1 | - | 1 | 70 | 103 | ||
DQ471861.1 | - | 1 | 63 | 103 | ||
DQ471864.1 | - | 1 | 121 | 103 | ||
DQ471865.1 | - | 1 | 121 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the DQ471858.1, DQ471859.1, DQ471860.1, DQ471861.1, DQ471864.1, DQ471865.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
