Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC099908.1 | - | 4 - 5 | 738 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000346618.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000535432.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | ENST00000613242.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001243076.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007138.hg.1 | NM_001949.4 | Refseq |
