Assay Details
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004699.3 | NP_004690.1 | 5 - 6 | 628 | 104 | |
GenBank mRNA | AD001530.1 | - | 5 - 6 | 593 | 104 | |
AK293119.1 | - | 5 - 6 | 582 | 104 | ||
AK313697.1 | - | 5 - 6 | 576 | 104 | ||
BC000028.2 | - | 5 - 6 | 599 | 3505365 | 104 | |
CR992165.1 | - | 5 - 6 | 628 | 104 | ||
D83260.1 | - | 5 - 6 | 498 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000158526.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000393600.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000464419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000478509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000481619.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000490480.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | ENST00000494278.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008833.hg.1 | NM_004699.3 | Refseq |
