Assay Details
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005911.5 | NP_005902.1 | 9 | 1994 | 114 | |
GenBank mRNA | BC001686.2 | - | 9 | 1791 | 3010820 | 114 |
BC001854.2 | - | 9 | 1791 | 3010820 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
