Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000612661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
ENST00000617290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009206.hg.1 |
NM_002356.6 |
Refseq |