Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000472116.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000474330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013435.hg.1 |
ENST00000487084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |