Assay Details
Gene Symbol: LINC01278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL080202.1 | - | 1 | 70 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000433061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000608623.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | ENST00000609401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011370.hg.1 | NR_015353.1 | Refseq |
