Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000586165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
ENST00000586165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000500968.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000586165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
ENST00000586165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009806.hg.1 |
NM_201286.3 |
Refseq |