Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000215631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000585359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000586759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000587887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000592937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
ENST00000593043.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006588.hg.1 |
NM_015675.3 |
Refseq |