Assay Details
Gene Symbol: ANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124230.1 | - | 1 | 1045 | 130 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK124230.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
